Print…

Anti-BCL7A Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB126753-100EA 0 EUR
BIRBORB126753-100 BIRBORB126753-200
Anti-BCL7A Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: BCL7A (B-cell CLL/lymphoma 7A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR