Print…

Anti-B4GALT1 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB126744-100EA 0 EUR
BIRBORB126744-100 BIRBORB126744-200
Anti-B4GALT1 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: B4GALT1 (UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR