Print…
Anti-B3GALT1 Rabbit Polyclonal Antibody
Fornecedor: Biorbyt
BIRBORB126743-200EA 0 EUR
BIRBORB126743-200 BIRBORB126743-100
Anti-B3GALT1 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: B3GALT1 (UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar