Print…

Anti-FBXL6 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18551-100EA 0 EUR
BIRBORB18551-100
Anti-FBXL6 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:8000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: FBXL6 (F-box and leucine-rich repeat protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR