Print…

Anti-FOXA1 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18549-20EA 0 EUR
BIRBORB18549-20 BIRBORB18549-100
Anti-FOXA1 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:8000; Western Blot: 0.2-0.6 ?g/ml

Type: Primary
Antigen: FOXA1 (forkhead box A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR