Print…

Anti-STAT3 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18540-100EA 0 EUR
BIRBORB18540-100
Anti-STAT3 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:4000

Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR