Print…

Anti-IDUA Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18500-100EA 0 EUR
BIRBORB18500-100
Anti-IDUA Goat Polyclonal Antibody
Anticorpos
ELISA: 1:128000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: IDUA (iduronidase, alpha-L-)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR