Print…

Anti-MURF2 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18497-100EA 0 EUR
BIRBORB18497-100
Anti-MURF2 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:16000; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: MURF2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR