Print…

Anti-CARD6 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18496-100EA 0 EUR
BIRBORB18496-100
Anti-CARD6 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:32000

Type: Primary
Antigen: CARD6 (caspase recruitment domain family, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR