Print…

Anti-RNF34 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18485-100EA 0 EUR
BIRBORB18485-100
Anti-RNF34 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:32000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: RNF34 (ring finger protein 34, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR