Print…

Anti-ANP32B Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18473-100EA 0 EUR
BIRBORB18473-100
Anti-ANP32B Goat Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: ANP32B (acidic (leucine-rich) nuclear phosphoprotein 32 family, member B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR