Print…

Anti-Peg12 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18465-100EA 0 EUR
BIRBORB18465-100
Anti-Peg12 Goat Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: Peg12
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR