Print…

Anti-NR2F6 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18427-100EA 0 EUR
BIRBORB18427-100
Anti-NR2F6 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:32000

Type: Primary
Antigen: NR2F6 (nuclear receptor subfamily 2, group F, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR