Print…
Anti-IRF5 Goat Polyclonal Antibody
Fornecedor: Biorbyt
BIRBORB18401-20EA 0 EUR
BIRBORB18401-20 BIRBORB18401-100
Anti-IRF5 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:32000; Western Blot: 0.1-0.3 ?g/ml
Type: Primary
Antigen: IRF5 (interferon regulatory factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar