Print…

Anti-ELMO3 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18361-100EA 0 EUR
BIRBORB18361-100
Anti-ELMO3 Goat Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: ELMO3 (engulfment and cell motility 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR