Print…

Anti-GRB2 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18345-20EA 0 EUR
BIRBORB18345-20 BIRBORB18345-100
Anti-GRB2 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:32000; Western Blot: 0.05-0.15 ?g/ml

Type: Primary
Antigen: GRB2 (growth factor receptor-bound protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR