Print…

Anti-NOS1 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18337-100EA 0 EUR
BIRBORB18337-100 BIRBORB18337-20
Anti-NOS1 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:32000; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: NOS1 (nitric oxide synthase 1 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR