Print…

Anti-DDAH2 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18336-100EA 0 EUR
BIRBORB18336-100 BIRBORB18336-20
Anti-DDAH2 Goat Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: DDAH2 (dimethylarginine dimethylaminohydrolase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR