Print…

Anti-PRAM1 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18326-100EA 0 EUR
BIRBORB18326-100
Anti-PRAM1 Goat Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: PRAM1 (PML-RARA regulated adaptor molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR