Print…

Anti-NFIL3 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18303-100EA 0 EUR
BIRBORB18303-100
Anti-NFIL3 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:8000; Western Blot: 0.5-1.5 ?g/ml

Type: Primary
Antigen: NFIL3 (nuclear factor, interleukin 3 regulated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR