Print…

Anti-NCF1 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18301-100EA 0 EUR
BIRBORB18301-100 BIRBORB18301-20
Anti-NCF1 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:20000; Western Blot: 0.5-1.5 ?g/ml

Type: Primary
Antigen: NCF1 (neutrophil cytosolic factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR