Print…
Anti-TRPM7 Goat Polyclonal Antibody
Fornecedor: Biorbyt
BIRBORB18297-100EA 0 EUR
BIRBORB18297-100 BIRBORB18297-20
Anti-TRPM7 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:8000
Type: Primary
Antigen: TRPM7 (transient receptor potential cation channel, subfamily M, member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar