Print…

Anti-DDB1 Goat Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18292-20EA 0 EUR
BIRBORB18292-20 BIRBORB18292-100
Anti-DDB1 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:32000; Western Blot: 0.5-1.5 ?g/ml

Type: Primary
Antigen: DDB1 (damage-specific DNA binding protein 1, 127kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR