Print…

Anti-TUBG1 Mouse Monoclonal Antibody (KHL)

Fornecedor: Biorbyt
BIRBORB18289-100EA 0 EUR
BIRBORB18289-100
Anti-TUBG1 Mouse Monoclonal Antibody (KHL)
Anticorpos
Type: Primary
Antigen: TUBG1 (tubulin, gamma 1)
Clonality: monoclonal
Clone:
Conjugation: KHL
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR