Print…

Anti-Smooth Muscle Actin Mouse Monoclonal Antibody

Fornecedor: Biorbyt
BIRBORB18282-100EA 0 EUR
BIRBORB18282-100
Anti-Smooth Muscle Actin Mouse Monoclonal Antibody
Anticorpos
Type: Primary
Antigen: Smooth Muscle Actin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR