Print…

Anti-Tropomyosin Mouse Monoclonal Antibody

Fornecedor: Biorbyt
BIRBORB18271-100EA 0 EUR
BIRBORB18271-100
Anti-Tropomyosin Mouse Monoclonal Antibody
Anticorpos
Type: Primary
Antigen: Tropomyosin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR