Print…

Anti-AKT1 Mouse Monoclonal Antibody KHL

Fornecedor: Biorbyt
BIRBORB18261-100EA 0 EUR
BIRBORB18261-100
Anti-AKT1 Mouse Monoclonal Antibody KHL
Anticorpos
Type: Primary
Antigen: AKT1 (v-akt murine thymoma viral oncogene homolog 1)
Clonality: monoclonal
Clone:
Conjugation: KHL
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR