Print…

Anti-PXN Mouse Monoclonal Antibody

Fornecedor: Biorbyt
BIRBORB18256-100EA 0 EUR
BIRBORB18256-100
Anti-PXN Mouse Monoclonal Antibody
Anticorpos
Type: Primary
Antigen: PXN (paxillin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR