Print…

Anti-p34cdc2 Mouse Monoclonal Antibody

Fornecedor: Biorbyt
BIRBORB18253-100EA 0 EUR
BIRBORB18253-100
Anti-p34cdc2 Mouse Monoclonal Antibody
Anticorpos
Type: Primary
Antigen: p34cdc2
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR