Print…

Anti-HSP90AA1 Mouse Monoclonal Antibody

Fornecedor: Biorbyt
BIRBORB18228-100EA 0 EUR
BIRBORB18228-100
Anti-HSP90AA1 Mouse Monoclonal Antibody
Anticorpos
Type: Primary
Antigen: HSP90AA1 (heat shock protein 90kDa alpha (cytosolic), class A member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR