Print…

Anti-BFSP1 Mouse monoclonal antibody

Fornecedor: Biorbyt
BIRBORB18218-100EA 0 EUR
BIRBORB18218-100
Anti-BFSP1 Mouse monoclonal antibody
Anticorpos
Type: Primary
Antigen: BFSP1 (beaded filament structural protein 1, filensin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR