Print…

Anti-MYC Mouse Monoclonal Antibody

Fornecedor: Biorbyt
BIRBORB18206-100EA 0 EUR
BIRBORB18206-100
Anti-MYC Mouse Monoclonal Antibody
Anticorpos
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR