Print…

Anti-CDK6 Mouse monoclonal antibody

Fornecedor: Biorbyt
BIRBORB18199-100EA 0 EUR
BIRBORB18199-100
Anti-CDK6 Mouse monoclonal antibody
Anticorpos
Type: Primary
Antigen: CDK6 (cyclin-dependent kinase 6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR