Print…

Anti-CALM1 Mouse Monoclonal Antibody

Fornecedor: Biorbyt
BIRBORB18188-100EA 0 EUR
BIRBORB18188-100
Anti-CALM1 Mouse Monoclonal Antibody
Anticorpos
Type: Primary
Antigen: CALM1 (calmodulin 1 (phosphorylase kinase, delta))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR