Print…

Anti-BIN1 Mouse Monoclonal Antibody

Fornecedor: Biorbyt
BIRBORB18183-100EA 0 EUR
BIRBORB18183-100
Anti-BIN1 Mouse Monoclonal Antibody
Anticorpos
Type: Primary
Antigen: BIN1 (bridging integrator 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR