Print…

Anti-BAP1 Mouse monoclonal antibody

Fornecedor: Biorbyt
BIRBORB18180-100EA 0 EUR
BIRBORB18180-100
Anti-BAP1 Mouse monoclonal antibody
Anticorpos
Type: Primary
Antigen: BAP1 (BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR