Print…

Anti-PPARG Rabbit polyclonal antibody

Fornecedor: Biorbyt
BIRBORB18174-100EA 344 EUR
BIRBORB18174-100
Anti-PPARG Rabbit polyclonal antibody
Anticorpos
Type: Primary
Antigen: PPARG (peroxisome proliferator-activated receptor gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR