Print…

Anti-MT-CO1 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18171-100EA 0 EUR
BIRBORB18171-100
Anti-MT-CO1 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: MT-CO1 (mitochondrially encoded cytochrome c oxidase I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR