Print…

Anti-SNAP25 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18169-100EA 0 EUR
BIRBORB18169-100
Anti-SNAP25 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: SNAP25 (synaptosomal-associated protein, 25kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR