Print…

Anti-Caspase-8 (P18) Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18162-100EA 0 EUR
BIRBORB18162-100
Anti-Caspase-8 (P18) Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: Caspase-8 (P18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR