Print…

Anti-TNFAIP1 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18159-100EA 0 EUR
BIRBORB18159-100
Anti-TNFAIP1 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: TNFAIP1 (tumor necrosis factor, alpha-induced protein 1 (endothelial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR