Print…

Anti-HPSE Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18129-100EA 0 EUR
BIRBORB18129-100
Anti-HPSE Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: HPSE (heparanase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR