Print…
Anti-ITGB2 Rabbit Polyclonal Antibody
Fornecedor: Biorbyt
BIRBORB18100-100EA 0 EUR
BIRBORB18100-100
Anti-ITGB2 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat