Print…
Anti-IFITM1 Rabbit Polyclonal Antibody
Fornecedor: Biorbyt
BIRBORB18088-100EA 0 EUR
BIRBORB18088-100
Anti-IFITM1 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: IFITM1 (interferon induced transmembrane protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human