Print…
Anti-STAT3 Rabbit Polyclonal Antibody
Fornecedor: Biorbyt
BIRBORB18086-100EA 0 EUR
BIRBORB18086-100
Anti-STAT3 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat