Print…
Anti-TIMP4 Rabbit Polyclonal Antibody
Fornecedor: Biorbyt
BIRBORB18075-100EA 0 EUR
BIRBORB18075-100
Anti-TIMP4 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: TIMP4 (TIMP metallopeptidase inhibitor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat