Print…
Anti-PMVK Rabbit Polyclonal Antibody
Fornecedor: Biorbyt
BIRBORB18064-100EA 0 EUR
BIRBORB18064-100
Anti-PMVK Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: PMVK (phosphomevalonate kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat