Print…

Anti-BNIP3 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18054-100EA 0 EUR
BIRBORB18054-100
Anti-BNIP3 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: BNIP3 (BCL2/adenovirus E1B 19kDa interacting protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR