Print…

Anti-LMNB1 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB18045-100EA 0 EUR
BIRBORB18045-100
Anti-LMNB1 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: LMNB1 (lamin B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR