Print…

Anti-BDNF Rabbit polyclonal antibody

Fornecedor: Biorbyt
BIRBORB18013-100EA 0 EUR
BIRBORB18013-100
Anti-BDNF Rabbit polyclonal antibody
Anticorpos
Type: Primary
Antigen: BDNF (brain-derived neurotrophic factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR